Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUMO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035458.1 | NP_001030535.1 | 1 - 2 | 111 | 58 | |
GenBank mRNA | BC102884.1 | - | 1 - 2 | 102 | 58 | |
BT020989.1 | - | 1 - 2 | 111 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |