Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035463.2 | NP_001030540.1 | 3 - 4 | 1174 | 153 | |
GenBank mRNA | BC103006.1 | - | 3 - 4 | 1175 | 153 | |
BC149598.1 | - | 3 - 4 | 1206 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |