Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035463.2 | NP_001030540.1 | 5 - 6 | 1433 | 62 | |
GenBank mRNA | BC103006.1 | - | 5 - 6 | 1434 | 62 | |
BC149598.1 | - | 5 - 6 | 1465 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |