Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035463.2 | NP_001030540.1 | 7 - 8 | 1720 | 83 | |
GenBank mRNA | BC103006.1 | - | 7 - 8 | 1721 | 83 | |
BC149598.1 | - | 7 - 8 | 1752 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Inventoried Probe spans exons |