Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035468.2 | NP_001030545.1 | 2 - 3 | 312 | 86 | |
GenBank mRNA | BC103235.1 | - | 2 - 3 | 313 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |