Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FXYD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037627.1 | NP_001032716.1 | 133 | 92 | ||
GenBank mRNA | BC103263.1 | - | 133 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Bos taurus |