Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOLLIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039961.1 | NP_001035050.1 | 1 - 2 | 91 | 129 | |
GenBank mRNA | BC123457.1 | - | 1 - 2 | 93 | 129 | |
BT021785.1 | - | 1 - 2 | 91 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |