Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOLLIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039961.1 | NP_001035050.1 | 5 - 6 | 679 | 95 | |
GenBank mRNA | BC123457.1 | - | 5 - 6 | 681 | 95 | |
BT021785.1 | - | 5 - 6 | 679 | 95 | ||
DQ319073.1 | - | 5 - 6 | 650 | 95 | ||
DQ407275.1 | - | 5 - 6 | 614 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |