Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COMMD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040597.1 | NP_001035687.1 | 168 | 70 | ||
GenBank mRNA | BC122799.1 | - | 3 - 4 | 188 | 70 | |
BT020731.1 | - | 3 - 4 | 153 | 70 | ||
BT020811.1 | - | 3 - 4 | 173 | 70 | ||
BT020814.1 | - | 3 - 4 | 159 | 70 | ||
BT020820.1 | - | 3 - 4 | 165 | 70 | ||
BT020878.1 | - | 3 - 4 | 171 | 70 | ||
BT020882.1 | - | 3 - 4 | 157 | 70 | ||
BT020897.1 | - | 3 - 4 | 160 | 70 | ||
BT020914.1 | - | 168 | 70 | |||
BT020930.1 | - | 3 - 4 | 160 | 70 | ||
BT020940.1 | - | 3 - 4 | 166 | 70 | ||
BT021643.1 | - | 3 - 4 | 162 | 70 | ||
BT021894.1 | - | 3 - 4 | 166 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |