Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POFUT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043342.2 | NP_001036807.1 | 1 - 2 | 197 | 65 | |
NM_001043343.1 | NP_001036808.1 | 1 - 2 | 199 | 65 | ||
NM_001043344.1 | NP_001036809.1 | 1 - 2 | 199 | 65 | ||
NM_001043345.2 | NP_001036810.1 | 1 - 2 | 197 | 65 | ||
NM_182986.2 | NP_892031.1 | 1 - 2 | 199 | 65 | ||
GenBank mRNA | AY344580.2 | - | 1 - 2 | 199 | 65 | |
AY344581.2 | - | 1 - 2 | 199 | 65 | ||
BC126762.1 | - | 1 - 2 | 172 | 65 | ||
BT021843.1 | - | 1 - 2 | 179 | 65 | ||
DQ138291.1 | - | 1 - 2 | 199 | 65 | ||
DQ138292.1 | - | 1 - 2 | 199 | 65 | ||
DQ138293.1 | - | 1 - 2 | 199 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |