Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075320.1 | NP_001068788.1 | 6 - 7 | 930 | 74 | |
GenBank mRNA | BT020787.1 | - | 6 - 7 | 930 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |