Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRCC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075790.1 | NP_001069258.1 | 7 - 8 | 567 | 82 | |
GenBank mRNA | BT021737.1 | - | 7 - 8 | 567 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Bos taurus Probe spans exons |