Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RUSF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076973.1 | NP_001070441.1 | 3 - 4 | 475 | 63 | |
GenBank mRNA | BT021776.1 | - | 3 - 4 | 475 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Bos taurus Probe spans exons |