Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRISP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079635.1 | NP_001073103.1 | 2 - 3 | 174 | 67 | |
GenBank mRNA | BC102058.1 | - | 2 - 3 | 174 | 67 | |
BT030687.1 | - | 2 - 3 | 183 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |