Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRISP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079635.1 | NP_001073103.1 | 6 - 7 | 629 | 109 | |
GenBank mRNA | BC102058.1 | - | 6 - 7 | 629 | 109 | |
BT030687.1 | - | 6 - 7 | 638 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |