Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080317.2 | NP_001073786.1 | 7 - 8 | 859 | 89 | |
GenBank mRNA | BC103047.1 | - | 7 - 8 | 860 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |