Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STOML3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080909.1 | NP_001074378.1 | 5 - 6 | 963 | 60 | |
GenBank mRNA | BT021609.1 | - | 5 - 6 | 963 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Bos taurus Probe spans exons |