Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173886.2 | NP_776311.2 | 4 - 5 | 720 | 69 | |
GenBank mRNA | EV611026.1 | - | 4 - 5 | 496 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Bos taurus Probe spans exons |