Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173968.3 | NP_776393.1 | 3 - 4 | 270 | 68 | |
GenBank mRNA | AF104105.1 | - | 3 - 4 | 267 | 68 | |
BC103415.1 | - | 3 - 4 | 270 | 68 | ||
BC120457.1 | - | 3 - 4 | 269 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Inventoried Probe spans exons |