Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174274.1 | NP_776699.1 | 3 - 4 | 1669 | 64 | |
GenBank mRNA | AF308815.2 | - | 2 - 3 | 1403 | 64 | |
AY027881.1 | - | 3 - 4 | 1708 | 64 | ||
AY027882.1 | - | 3 - 4 | 1708 | 64 | ||
BC151599.1 | - | 3 - 4 | 1617 | 64 | ||
BT025444.1 | - | 3 - 4 | 1663 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |