Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174274.1 | NP_776699.1 | 5 - 6 | 1935 | 93 | |
GenBank mRNA | AF308815.2 | - | 4 - 5 | 1669 | 93 | |
AY027881.1 | - | 5 - 6 | 1974 | 93 | ||
AY027882.1 | - | 5 - 6 | 1974 | 93 | ||
BC151599.1 | - | 5 - 6 | 1883 | 93 | ||
BT025444.1 | - | 5 - 6 | 1929 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |