Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PROKR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174330.1 | NP_776755.1 | 1 - 2 | 480 | 76 | |
GenBank mRNA | AY089972.1 | - | 1 - 2 | 480 | 76 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Recommended Assay Bos taurus Probe spans exons |