Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCGR1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174538.2 | NP_776963.1 | 4 - 5 | 583 | 100 | |
GenBank mRNA | AF162866.1 | - | 4 - 5 | 583 | 100 | |
BC112691.1 | - | 4 - 5 | 595 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |