Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC1A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174601.2 | NP_777026.1 | 3 - 4 | 1273 | 97 | |
GenBank mRNA | AY039236.1 | - | 3 - 4 | 1273 | 97 | |
BC123803.1 | - | 3 - 4 | 862 | 97 | ||
LC005614.1 | - | 3 - 4 | 658 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |