Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD11B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174642.2 | NP_777067.1 | 2 - 3 | 574 | 107 | |
GenBank mRNA | AF074706.2 | - | 2 - 3 | 576 | 107 | |
BC102488.1 | - | 2 - 3 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |