Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLCO1A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174654.2 | NP_777079.1 | 4 - 5 | 603 | 116 | |
GenBank mRNA | AJ508718.1 | - | 4 - 5 | 603 | 116 | |
AY052775.1 | - | 5 - 6 | 687 | 116 | ||
BC133437.1 | - | 4 - 5 | 648 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |