Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHIA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174699.2 | NP_777124.1 | 3 - 4 | 272 | 100 | |
GenBank mRNA | AB051629.1 | - | 3 - 4 | 272 | 100 | |
BC102931.1 | - | 3 - 4 | 272 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |