Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHIA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174699.2 | NP_777124.1 | 9 - 10 | 1044 | 78 | |
GenBank mRNA | AB051629.1 | - | 9 - 10 | 1044 | 78 | |
BC102931.1 | - | 9 - 10 | 1044 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |