Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB27B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174738.2 | NP_777163.1 | 5 - 6 | 793 | 58 | |
GenBank mRNA | AY010899.1 | - | 5 - 6 | 793 | 58 | |
BC122732.1 | - | 6 - 7 | 638 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |