Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FSCN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176633.3 | NP_788806.1 | 4 - 5 | 1302 | 73 | |
GenBank mRNA | AB003485.1 | - | 4 - 5 | 1304 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Bos taurus Probe spans exons |