Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176640.2 | NP_788813.1 | 3 - 4 | 432 | 78 | |
GenBank mRNA | AJ440723.1 | - | 3 - 4 | 439 | 78 | |
BC112444.1 | - | 3 - 4 | 438 | 78 | ||
BT021802.1 | - | 3 - 4 | 432 | 78 | ||
BT026188.1 | - | 3 - 4 | 423 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |