Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADGRE5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176661.1 | NP_788834.1 | 9 - 10 | 922 | 116 | |
GenBank mRNA | AJ416058.1 | - | 9 - 10 | 922 | 116 | |
AJ627191.1 | - | 11 - 12 | 1253 | 116 | ||
BC149039.1 | - | 9 - 10 | 973 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Bos taurus Probe spans exons |