Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFA13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176672.2 | NP_788845.1 | 3 - 4 | 276 | 67 | |
GenBank mRNA | AJ316011.1 | - | 3 - 4 | 276 | 67 | |
BC102406.1 | - | 3 - 4 | 261 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |