Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NNAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178323.3 | NP_847893.1 | 1 - 2 | 167 | 76 | |
GenBank mRNA | AB047030.1 | - | 1 - 2 | 155 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Bos taurus Probe spans exons |