Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178572.2 | NP_848667.1 | 4 - 5 | 493 | 71 | |
GenBank mRNA | AY240020.1 | - | 4 - 5 | 493 | 71 | |
BC103260.1 | - | 4 - 5 | 547 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |