Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178574.3 | NP_848669.1 | 8 - 9 | 990 | 61 | |
GenBank mRNA | AY219899.1 | - | 8 - 9 | 991 | 61 | |
BC102160.1 | - | 8 - 9 | 961 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |