Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKIG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205812.1 | NP_991381.1 | 3 - 4 | 351 | 67 | |
GenBank mRNA | AY339896.1 | - | 3 - 4 | 351 | 67 | |
BC120316.1 | - | 3 - 4 | 323 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |