Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015556.2 | NP_001015556.1 | 2 | 323 | 102 | |
GenBank mRNA | BC104507.1 | - | 2 | 323 | 102 | |
BT021047.1 | - | 4 - 5 | 322 | 102 | ||
DQ403039.1 | - | 3 - 4 | 231 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Both primers and probe map within a single exon |