Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025341.3 | NP_001020512.1 | 128 | 109 | ||
GenBank mRNA | AY911357.1 | - | 2 - 3 | 130 | 109 | |
BC102125.1 | - | 131 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |