Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025342.2 | NP_001020513.1 | 1 | 340 | 91 | |
GenBank mRNA | AY911350.1 | - | 4 - 5 | 343 | 91 | |
BC102749.1 | - | 1 | 340 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Both primers and probe map within a single exon |