Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033610.1 | NP_001028782.1 | 3 - 4 | 697 | 78 | |
GenBank mRNA | BC103339.1 | - | 3 - 4 | 697 | 78 | |
K03532.1 | - | 3 - 4 | 291 | 78 | ||
X12877.1 | - | 291 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Inventoried Probe spans exons |