Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033626.1 | NP_001028798.1 | 1 - 2 | 115 | 64 | |
GenBank mRNA | BC102564.1 | - | 1 - 2 | 115 | 64 | |
BC142072.1 | - | 1 - 2 | 113 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |