Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBL4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034214.2 | NP_001029386.1 | 1 | 97 | 100 | |
GenBank mRNA | BC103189.1 | - | 1 | 98 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Both primers and probe map within a single exon |