Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CENPH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034236.2 | NP_001029408.1 | 7 - 8 | 564 | 120 | |
GenBank mRNA | BC102350.1 | - | 7 - 8 | 565 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |