Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANAPC15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034243.1 | NP_001029415.1 | 2 - 3 | 165 | 65 | |
GenBank mRNA | BC103197.1 | - | 2 - 3 | 165 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |