Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CES2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034260.2 | NP_001029432.1 | 7 - 8 | 1165 | 87 | |
GenBank mRNA | BC102288.1 | - | 7 - 8 | 1166 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |