Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTSF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034273.2 | NP_001029445.1 | 6 - 7 | 525 | 113 | |
GenBank mRNA | BC102713.1 | - | 6 - 7 | 526 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |