Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ECSIT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034286.1 | NP_001029458.1 | 6 - 7 | 1027 | 83 | |
GenBank mRNA | BC104572.1 | - | 6 - 7 | 1027 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |