Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ETHE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034344.1 | NP_001029516.1 | 6 - 7 | 753 | 69 | |
GenBank mRNA | BC102496.1 | - | 6 - 7 | 753 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |