Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFRP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034393.1 | NP_001029565.1 | 1 - 2 | 742 | 101 | |
GenBank mRNA | BC104536.1 | - | 1 - 2 | 742 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Recommended Assay Bos taurus Inventoried Probe spans exons |